Skip to Content
Merck
All Photos(1)

Documents

475734

Sigma-Aldrich

Glycerol diglycidyl ether

technical grade

Synonym(s):

Glycerin diglycidyl ether, Glycerol diglycidyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O5
CAS Number:
Molecular Weight:
204.22
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

refractive index

n20/D 1.482 (lit.)

density

1.229 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCC(COCC1CO1)OCC2CO2.OC(COCC3CO3)COCC4CO4

InChI

1S/2C9H16O5/c10-7(1-11-3-8-5-13-8)2-12-4-9-6-14-9;10-1-7(12-5-9-6-14-9)2-11-3-8-4-13-8/h2*7-10H,1-6H2

InChI key

QFMWJDGCJROGRC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycerol diglycidyl ether (GDE) is an aliphatic epoxy monomer that can be used as a diepoxy crosslinker. It has a low viscous epoxy matrix that can be cured with aliphatic amines to form a thermosetting material with good flexibility. Its properties include low shrinkage, good adhesion, and good thermo-mechanical properties.

Application

GDE can be used in the formation of epoxy materials, which can further be used in electrical instruments and biodegradable plastics.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The relationship between composition and electrical properties of brass covered by a nanocomposite coating of glycerol diglycidyl ether: An EIS and DMTA study
Omrani A, et al.
Progress in Organic Coatings, 76(2-3), 360-366 (2013)
Arpad A Vass
Forensic science international, 301, 149-159 (2019-06-04)
This case report summarises the investigation of a death scene in the trunk of a car. Air sampling, laser-induced breakdown spectroscopy, and gas chromatography/mass spectrometry on samples of carpet and tyre well scrapings from the vehicle's trunk were utilised to
Saif urRehman et al.
Colloids and surfaces. B, Biointerfaces, 136, 1156-1165 (2015-11-29)
Here, we report a new microgel preparation from tris(2-aminoethyl)amine (TAEA) and glycerol diglycidyl ether (GDE) as p(TAEA-co-GDE) via simple microemulsion polymerization/crosslinking by using L-α lecithin as surfactant and gasoline as organic phase. The p(TAEA-co-GDE) microgels were visualized using optical microscopy
Fabrication and characterization of chemically crosslinked keratin films
Tanabe T, et al.
Materials Science and Engineering, C, 24(3), 441-446 (2004)
Photocross-linking of glycerol diglycidyl ether with reactive diluents
Kasetaite S, et al.
Polymer Bull., 72(12), 3191-3208 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service