Skip to Content
Merck
All Photos(1)

Key Documents

467979

Sigma-Aldrich

1,3-Dithiolane

97%

Synonym(s):

1,3-Dithiacyclopentane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6S2
CAS Number:
Molecular Weight:
106.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.599 (lit.)

bp

183 °C (lit.)

density

1.235 g/mL at 25 °C (lit.)

functional group

thioether

SMILES string

C1CSCS1

InChI

1S/C3H6S2/c1-2-5-3-4-1/h1-3H2

InChI key

IMLSAISZLJGWPP-UHFFFAOYSA-N

General description

1,3-Dithiolane ,a sulfur containing heterocyclic building block, is a cyclic thioether. Fragmentation modes of 1,3-oxathiolane under electron-impact have been investigated.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Papanikos et al.
Journal of combinatorial chemistry, 6(2), 181-195 (2004-03-09)
A synthetic strategy for the formation of resin-bound internal alpha-keto amide peptides suitable for protease inhibitor screening on solid support is presented. This general approach is based on the incorporation of alpha-keto amide building blocks during solid-phase peptide synthesis (SPPS).
Sylvie Goncalves et al.
Chemical communications (Cambridge, England), 46(31), 5778-5780 (2010-07-06)
A concise and diastereoselective formal total synthesis of triptolide, a natural product with a wide range of biological properties, is described. The key reaction is an unprecedented 6-endo-Trig cationic cyclization of a 2-alkenyl-1,3-dithiolane precursor induced by TMSOTf as Lewis acid.
I M Hussaini et al.
Acta pharmacologica Sinica, 21(10), 897-904 (2001-08-15)
To investigate the effect of a group of novel synthetic dithiolane analogs of lignans and a well characterized platelet-activating factor (PAF) receptor antagonist, L659,989 on PAF-receptor binding, IFN-gamma- and lipopolysaccharide (LPS)-induced NO production, and steady-state inducible nitric-oxide synthase (iNOS) mRNA
Differentiation between carbonyls and acetals in 1, 3-dithiane and 1, 3-dithiolane synthesis catalyzed by organotin triflates.
Sato T, et al.
The Journal of Organic Chemistry, 58(18), 4971-4978 (1993)
Ionization and dissociation of cyclic ethers and thioethers by electron-impact. A comparison between 1, 3-dioxolane, 1, 3-dithiolane and 1, 3-oxathiolane.
Conde-Caprace G and Collin JE.
Org. Mass Spectrom., 6(4), 415-423 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service