Skip to Content
Merck
All Photos(2)

Documents

458821

Sigma-Aldrich

2-Chloropyridine-3-carbonyl chloride

98%

Synonym(s):

2-Chloronicotinoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl2NO
CAS Number:
Molecular Weight:
176.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

39-44 °C (lit.)

SMILES string

ClC(=O)c1cccnc1Cl

InChI

1S/C6H3Cl2NO/c7-5-4(6(8)10)2-1-3-9-5/h1-3H

InChI key

RXTRRIFWCJEMEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and biological evaluation of new benzothiazoles as antimicrobial agents.
Al-Talib M, et al.
Arabian Journal of Chemistry (2011)
Chemistry of heterocyclic compounds. 54. Nicotinic acid crown ethers. Synthesis of macrocyclic lactones from 2-chloronicotinic acid and polyethylene glycols. Template effect on the cyclization.
Newkome GR, et al.
The Journal of Organic Chemistry, 45(4), 626-628 (1980)
Yu-Peng Hua et al.
Acta crystallographica. Section E, Structure reports online, 65(Pt 2), o373-o373 (2009-01-01)
In the crystal structure of the title compound, C(6)H(5)ClN(2)O, the dihedral angle between the pyridine ring and the carboxamine group is 63.88 (8)°. Inter-molecular N-H⋯N and N-H⋯O hydrogen bonds link the mol-ecules into a two-dimensional network.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service