Skip to Content
Merck
All Photos(2)

Documents

437646

Sigma-Aldrich

Sodium hydrogen difluoride

granular, 98%

Synonym(s):

Sodium acid fluoride, Sodium bifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHF2
CAS Number:
Molecular Weight:
61.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

granular

SMILES string

[F-].[Na+].F[H]

InChI

1S/2FH.Na/h2*1H;/q;;+1/p-1

InChI key

BFXAWOHHDUIALU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Employed recently in the room temperature synthesis of fluoride glasses. Starting material for solid state preparation of Na3MnF6 for further high pressure structural determination studies.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Journal of Non-Crystalline Solids, 184, 166-166 (1995)
Carlson, S. et al.
Inorganic Chemistry, 37, 1486-1486 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service