Skip to Content
Merck
All Photos(1)

Key Documents

433985

Sigma-Aldrich

5-Bromo-2-furaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3BrO2
CAS Number:
Molecular Weight:
174.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

bp

112 °C/16 mmHg (lit.)

mp

82-85 °C (lit.)

storage temp.

2-8°C

SMILES string

Brc1ccc(C=O)o1

InChI

1S/C5H3BrO2/c6-5-2-1-4(3-7)8-5/h1-3H

InChI key

WJTFHWXMITZNHS-UHFFFAOYSA-N

Application

5-Bromo-2-furaldehyde may be employed for the following syntheses:
  • 5-substituted 2-furaldehydes
  • anilines, through a novel one-pot, two-step amination/Diels-Alder procedure
  • 5-(5′,8′-dimethyl-9′-tert-butoxycarbonyl-9′H-carbazol-3′-yl)-furan-2-carbaldehyde
  • 5-(6-hydroxyhexyl)-2-furaldehyde
  • 5-methylsulfonyl-2-furaldehyde
  • 5-phenylsulfonyl-2-furaldehyde
  • 5-(4-acetamidobenzylsulfonyl)-2-furaldehyde
  • 5-iodo-2-furaldehyde

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raouf Medimagh et al.
The Journal of organic chemistry, 73(6), 2191-2197 (2008-02-28)
Selective metal-free amination and Diels-Alder reactions are described in the furan series, leading to polysubstituted anilines or to stable oxabicyclic adducts in high yield. Interestingly, anilines are conveniently prepared through a novel one-pot, two-step amination/Diels-Alder procedure from commercially available 5-bromo-2-furaldehyde.
Aminomethylations via cross-coupling of potassium organotrifluoroborates with aryl bromides.
Molander GA and Sandrock DL.
Organic Letters, 9(8), 1597-1600 (2007)
Furan derivatives. LXXXV11. The synthesis and ultraviolet spectra of 5-(4-X-phenyIsulfonyI)-2-furaldehydes and 2-cyano-3-[5-(4-X-phenyl-sulfonyl)-2-furyl] acrylonitriles.
Kada R and Kovac J.
Chemical Papers, 30(4), 502-507 (1976)
A synthesis strategy yielding skeletally diverse small molecules combinatorially.
Burke MD, et al.
Journal of the American Chemical Society, 126(43), 14095-14104 (2004)
Efficient and Simple Synthesis of 6-Aryl-1,4-dimethyl-9H-carbazoles.
Caruso A, et al.
Molecules (Basel), 13(6), 1312-1320 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service