Skip to Content
Merck
All Photos(1)

Documents

416762

Sigma-Aldrich

5-Bromo-2,4-dichloropyrimidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4HBrCl2N2
CAS Number:
Molecular Weight:
227.87
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

refractive index

n20/D 1.603 (lit.)

bp

128 °C/15 mmHg (lit.)

mp

29-30 °C (lit.)

density

1.781 g/mL at 25 °C (lit.)

SMILES string

Clc1ncc(Br)c(Cl)n1

InChI

1S/C4HBrCl2N2/c5-2-1-8-4(7)9-3(2)6/h1H

InChI key

SIKXIUWKPGWBBF-UHFFFAOYSA-N

General description

5-Bromo-2,4-dichloropyrimidine is a halogenoprimidine. It has been reported as an intermediate during the synthesis of 2,4-di-t-butoxy-5-bromopyrimidine and 2,4-di-t-butoxy-5-pyrimidineboronic acid.

Application

5-Bromo-2,4-dichloropyrimidine may be employed as starting reagent for the synthesis of positive allosteric modulators for GABAB receptors (drug-like class of compounds) and pyridinepyrimidine analogs.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sébastien Guery et al.
Bioorganic & medicinal chemistry letters, 17(22), 6206-6211 (2007-09-22)
The optimization of GS39783 into potent, selective, and safe positive allosteric modulators of GABA(B) receptors is presented.
Synthesis of Various 5-Substituted Uracils.
Peters D, et al.
Journal of Heterocyclic Chemistry, 27(7), 2165-2170 (1990)
Pyrimidines. Part VIII. Halogeno-and hydrazino-pyrimidines.
Chesterfield J, et al.
Journal of the Chemical Society, 3478-3481 (1955)
Synthesis, 861-861 (2006)
Synthesis, 5, 728-734 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service