Skip to Content
Merck
All Photos(1)

Documents

408352

Sigma-Aldrich

Poly(propylene glycol) methacrylate

average Mn ~375

Synonym(s):

Polypropylene glycol monomethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

mol wt

average Mn ~375

contains

800 ppm MEHQ as inhibitor

refractive index

n20/D 1.449

density

1.01 g/mL at 25 °C (lit.)

SMILES string

CC(O)CO.CC(=C)C(O)=O

InChI

1S/C7H12O3/c1-6(2)7(9)10-5-3-4-8/h8H,1,3-5H2,2H3

InChI key

GNSFRPWPOGYVLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service