Skip to Content
Merck
All Photos(2)

Key Documents

399981

Sigma-Aldrich

4,4′-(9-Fluorenylidene)diphenol

97%

Synonym(s):

9,9-Bis(4-hydroxyphenyl)fluorene, BPFL, BisP-FL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H18O2
CAS Number:
Molecular Weight:
350.41
Beilstein:
2060986
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

224-226 °C (lit.)

SMILES string

Oc1ccc(cc1)C3(c2ccc(O)cc2)c4ccccc4-c5ccccc35

InChI

1S/C25H18O2/c26-19-13-9-17(10-14-19)25(18-11-15-20(27)16-12-18)23-7-3-1-5-21(23)22-6-2-4-8-24(22)25/h1-16,26-27H

InChI key

YWFPGFJLYRKYJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the synthesis of novel poly(arylene ethers).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Journal of Polymer Science Part A: Polymer Chemistry, 29, 1045-1045 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service