Skip to Content
Merck
All Photos(1)

Documents

378399

Sigma-Aldrich

Silicone oil

viscosity 1,000 cSt (25 °C)

Synonym(s):

Dimethylpolysiloxane, Dimethylsilcone fluid, Poly(dimethylsiloxane)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-Si(CH3)2O-]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

vapor density

>1 (vs air)

vapor pressure

<5 mmHg ( 25 °C)
5 mmHg ( 20 °C)

form

viscous liquid

refractive index

n20/D 1.403 (lit.)

viscosity

1,000 cSt(25 °C)

bp

>140 °C/0.002 mmHg (lit.)

density

0.97 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Silicone oil is a liquid based siloxane that is part of the methyl silicone fluid system. It has a viscosity of 1000 cSt with a refractive index of ~ 1.403 and a dielectric strength of ~ 14 kV/mm. It′s surface tension tends to increase with an increase in the viscosity.

Application

Silicone oil can be used for a wide range of applications such as: heat transferring medium in chemical and petrochemical industries, a dielectric coolant, protective coatings for building materials, a cosmetic additive.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

214.0 °F - closed cup

Flash Point(C)

101.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silicones
Moretto H, et al.
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Recent developments and applications of protective silicone coatings: A review of PDMS functional materials
Eduok U, et al.
Progress in Organic Coatings, 111, 124-163 (2017)
Mechanism of Stabilization of Silicone Oil- Water Emulsions Using Hybrid Siloxane Polymers
Mehta SC and Somasundaran P
Langmuir, 24(9), 4558-4563 (2008)
Nobuo Yoshimoto et al.
Scientific reports, 3, 1191-1191 (2013-02-05)
When establishing the most appropriate cells from the huge numbers of a cell library for practical use of cells in regenerative medicine and production of various biopharmaceuticals, cell heterogeneity often found in an isogenic cell population limits the refinement of
Bo Xian et al.
Aging cell, 12(3), 398-409 (2013-02-28)
Caenorhabditis elegans is a leading model organism for studying the basic mechanisms of aging. Progress has been limited, however, by the lack of an automated system for quantitative analysis of longevity and mean lifespan. To address this barrier, we developed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service