Skip to Content
Merck
All Photos(2)

Key Documents

373613

Sigma-Aldrich

2,2-Dimethylcyclohexanone

92%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H8(=O)
CAS Number:
Molecular Weight:
126.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

92%

refractive index

n20/D 1.448 (lit.)

bp

169-170 °C/768 mmHg (lit.)

mp

−20 °C (lit.)

density

0.912 g/mL at 25 °C (lit.)

SMILES string

CC1(C)CCCCC1=O

InChI

1S/C8H14O/c1-8(2)6-4-3-5-7(8)9/h3-6H2,1-2H3

InChI key

KNSPBSQWRKKAPI-UHFFFAOYSA-N

General description

2,2-Dimethylcyclohexanone is a sterically hindered ketone. Mg-TiCl4-catalyzed CH2-transfer reaction of 2,2-dimethylcyclohexanone with CH2Cl2 is reported.

Application

2,2-Dimethylcyclohexanone may be used in the preparation of 6,6-dimethyl-1-vinylcyclohexene.

Other Notes

Remainder 2-methylcyclohexanone

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and antioxidant activity of rosmariquinone and several analogues.
Hall CA, et al.
Journal of Agricultural and Food Chemistry, 46(4), 1303-1310 (1998)
Tu-Hsin Yan et al.
Organic letters, 6(26), 4961-4963 (2004-12-21)
[reaction: see text] This Mg-TiCl4-promoted CH2-transfer reaction of CH2Cl2 represents an extremely simple, practical, and efficient methylenation of a variety of ketones and aldehydes, especially in enolizable or sterically hindered ketones such as 2,2-dimethylcyclohexanone, camphor, and fenchone.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service