Skip to Content
Merck
All Photos(1)

Key Documents

367672

Sigma-Aldrich

4,4′-(Hexafluoroisopropylidene)bis(benzoic acid)

98%

Synonym(s):

2,2-Bis(4-carboxylphenyl)-1,1,1,3,3,3-hexafluoropropane, 2,2-Bis(p-carboxyphenyl)hexafluoropropane, 4,4′-(1,1,1,3,3,3-Hexafluoropropane-2,2-diyl)dibenzoic acid, 4,4′-Hexafluoroisopropylidenediphenyl-1,1′-dicarboxylic acid, 4,4′-[2,2,2-Trifluoro-1-(trifluoromethyl)ethylidene]bis[benzoic acid], Hexafluoro-2,2-bis(4-carboxyphenyl)propane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C(C6H4CO2H)2
CAS Number:
Molecular Weight:
392.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

272-274 °C (lit.)

SMILES string

OC(=O)c1ccc(cc1)C(c2ccc(cc2)C(O)=O)(C(F)(F)F)C(F)(F)F

InChI

1S/C17H10F6O4/c18-16(19,20)15(17(21,22)23,11-5-1-9(2-6-11)13(24)25)12-7-3-10(4-8-12)14(26)27/h1-8H,(H,24,25)(H,26,27)

InChI key

PHQYMDAUTAXXFZ-UHFFFAOYSA-N

General description

4,4′-(Hexafluoroisopropylidene)bis(benzoic acid) is an aromatic dicarboxylic acid. It is a versatile ligand.

Application

4,4′-(Hexafluoroisopropylidene)bis(benzoic acid) is suitable for use as monomer in the synthesis of polyoxadiazole and polytriazole. It may be used:
  • in the solvothermal synthesis of new coordination compounds with different transition metal ions
  • as bent ligand in the solvothermal synthesis of metal-organic coordination polymers
  • in the synthesis of RPF-4, a family of polymeric frameworks

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheida Esmaielzadeh et al.
Ultrasonics sonochemistry, 41, 641-650 (2017-11-16)
Novel sulfonated polybenzimidazole (s-PBI)/cellulose/silica bionanocomposite membranes were prepared from fluorine-containing s-PBI copolymer with a cellulose/silica precursor and a bonding agent. The introduction of the bonding agent results in the reinforcing interfacial interaction between s-PBI chains and the cellulose/silica nanoparticles. Commercially
Synthesis and fabrication of nanostructured hydrophobic polyazole membranes for low-energy water recovery.
Maab H, et al.
Journal of Membrane Science , 423, 11-19 (2012)
Wenbin Yang et al.
Inorganic chemistry, 48(23), 11067-11078 (2009-12-01)
We have synthesized five new metal-organic coordination polymers incorporating the bent ligand H(2)hfipbb [4,4'-(hexafluoroisopropylidene)bis(benzoic acid)] with different transition metal ions and co-ligands via solvothermal reactions to give [Zn(2)(hfipbb)(2)(py)(2)] x DMF (1), [Zn(2)(hfipbb)(2)(4,4'-bipy)(H(2)O)] (2), [Zn(2)(hfipbb)(2)(bpdab)].2DMF (3), [Cd(2)(hfipbb)(2)(DMF)(2)] x 2 DMF (4)
Changchun Ji et al.
Dalton transactions (Cambridge, England : 2003), 39(35), 8240-8247 (2010-08-05)
Four new coordination compounds, namely {[Cd(3)(L)(2)(mu(3)-OH)(2)(H(2)O)] x H(2)O}(n) (1), {Ni(L)(bipy)}(n) (2), {Cu(3)(L)(2)(bipy)(mu(2)-OH)(2)(DMF)(2)}(n) (3), and {Co(2)L(2)(H(2)L)(H(2)O)}(n) (4), have been synthesized by the solvothermal reaction of 4,4'-(hexafluoroisopropylidene)bis(benzoic acid) (H(2)L) with different transition metal ions in the presence of co-ligand 4,4'-bipyridine (bipy). Compound
Felipe Gándara et al.
Inorganic chemistry, 48(11), 4707-4713 (2009-04-21)
RPF-4 is a family of polymeric frameworks prepared with rare-earth elements and the versatile ligand 4,4'-(hexafluoroisopropylidene)bis(benzoic acid). We have found that during the synthesis procedure up to three different polymorphs can be obtained. Their crystal structures are here presented. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service