Skip to Content
Merck
All Photos(1)

Key Documents

33820

Sigma-Aldrich

Dibenzyl sulfide

≥95.0% (HPLC)

Synonym(s):

Benzyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2S
CAS Number:
Molecular Weight:
214.33
Beilstein:
1911157
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (HPLC)

form

solid

mp

44-47 °C (lit.)

functional group

phenyl
thioether

SMILES string

C(SCc1ccccc1)c2ccccc2

InChI

1S/C14H14S/c1-3-7-13(8-4-1)11-15-12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

LUFPJJNWMYZRQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfur K-edge XANES spectra of dibenzyl sulfide has been measured. Adsorption modes of dibenzyl disulphide on bare and iron-coated silver in 0.5M HCl was studied by surface-enhanced Raman scattering (SERS) spectroscopy. Dibenzyl sulfide undergoes oxidation to dibenzyl sulfoxide by SiO2-HNO3/KBr.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Francesco Naso et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(48), 13417-13426 (2009-11-13)
An experimental investigation of the enantioselective oxidation of aryl benzyl sulfides by tert-butyl hydroperoxide in the presence of a titanium/hydrobenzoin catalyst has shown that these sulfides are ideal substrates for this catalytic system, with negligible interference by the substituents on
I L Genz et al.
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten und Hygiene. Zweite naturwissenschaftliche Abteilung: Mikrobiologie der Landwirtschaft der Technologie und des Umweltschutzes, 135(4), 308-312 (1980-01-01)
A diffusion cell type which is suitable for investigation of water-insoluble compounds is shown. The chamber consists of two compartments, the so-called bottom-chamber, which contains the solid substance under investigation, and the main chamber, which is above it. In this
N Itoh et al.
FEBS letters, 412(2), 281-284 (1997-07-28)
The degradation of thiodiglycol (I) and benzyl sulfide (II) was attempted using Coriolus versicolor and Tyromyces palustris to investigate the potential ability of basidiomycetes to degrade Yperite (bis(2-chloroethyl) sulfide), a mass-produced and stored chemical warfare agent. I was very rapidly
Jonathan D Van Hamme et al.
Applied and environmental microbiology, 69(2), 1320-1324 (2003-02-07)
Microbial metabolism of organosulfur compounds is of interest in the petroleum industry for in-field viscosity reduction and desulfurization. Here, dibenzyl sulfide (DBS) metabolism in white rot fungi was studied. Trametes trogii UAMH 8156, Trametes hirsuta UAMH 8165, Phanerochaete chrysosporium ATCC
Robert Beck et al.
Dalton transactions (Cambridge, England : 2003), (37)(37), 4981-4983 (2008-09-20)
C-H activation through the coordination of a benzyl sulfide anchoring group with a C-S bond cleavage at a Co(v) center constitutes a regiospecific access to four- and five-membered metallacycles under mild conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service