Skip to Content
Merck
All Photos(3)

Key Documents

319740

Sigma-Aldrich

2-Nitrofuran

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3NO3
CAS Number:
Molecular Weight:
113.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

crystals

bp

84 °C/13 mmHg (lit.)

mp

29-33 °C (lit.)

functional group

nitro

SMILES string

[O-][N+](=O)c1ccco1

InChI

1S/C4H3NO3/c6-5(7)4-2-1-3-8-4/h1-3H

InChI key

FUBFWTUFPGFHOJ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liyun Liu et al.
Frontiers in cellular and infection microbiology, 8, 233-233 (2018-07-28)
Objectives:Citrobacter freundii is a frequent cause of nosocomial infections and a known cause of diarrheal infections, and has increasingly become multidrug resistant (MDR). In this study, we aimed to determine the genetic diversity, the antimicrobial resistance profiles and in vitro
Pierre Guichard et al.
Food chemistry, 342, 128389-128389 (2020-12-04)
LC-MS/MS method for confirmation of nitrofuran metabolites in meat and aquaculture products, including the nifursol metabolite (DNSH), was developed. The nitrofuran metabolites investigated were as follows: 3-amino-2-oxazolidinone (AOZ), 3-amino-5-morpholinomethyl-2-oxazolidinone (AMOZ), 1-aminohydantoine (AHD), semicarbazide (SEM) and 3,5-dinitrosalicylic acid hydrazide (DNSH). The
Jinyan Gong et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1146, 122018-122018 (2020-04-26)
This study established a validated analytical method for the first time on the determination of nitrofuran metabolites, including semicarbazide (SEM), 1-aminohydantoin (AHD), 3-amino-2-oxazolidinone (AOZ) and 3-amino-5-morpholinomethyl-2-oxazolinone (AMOZ) in gelatin Chinese medicine. A C18 column with the mobile phase consisting of
Brian T Veach et al.
Journal of AOAC International, 101(3), 897-904 (2017-12-01)
This paper describes a rapid and robust method utilizing a single liquid-liquid extraction for the quantitation and confirmation of chloramphenicol, florfenicol, and nitrofuran metabolites in honey. This methodology combines two previous extraction methods into a single extraction procedure and utilizes
Hui Li et al.
Xenobiotica; the fate of foreign compounds in biological systems, 47(2), 103-111 (2016-04-20)
1. Nitrofurantoin (NFT), a 5-nitrofuran derivative, has been widely used for the treatment of specific urinary tract infections. It has been reported that exposure to NFT was associated with various adverse effects, particularly hepatotoxicity and pneumotoxicity. The objective of the study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service