Skip to Content
Merck
All Photos(3)

Key Documents

278394

Sigma-Aldrich

3,5-Dimethyl-1-hexyn-3-ol

98%

Synonym(s):

3,5-Dimethyl-1-hexyne-3-ol, 3,5-Dimethylhexyn-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)(OH)C≡CH
CAS Number:
Molecular Weight:
126.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.34 (vs air)

vapor pressure

4.5 mmHg ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

150-151 °C (lit.)

density

0.859 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC(C)(O)C#C

InChI

1S/C8H14O/c1-5-8(4,9)6-7(2)3/h1,7,9H,6H2,2-4H3

InChI key

NECRQCBKTGZNMH-UHFFFAOYSA-N

General description

Bimolecular rate constant for the reaction of the hydroxyl radical (OH) with 3,5-dimethyl-1-hexyn-3-ol has been been measured using the relative rate technique. Reaction of CO2 with 3,5-dimethyl-1-hexyn-3-ol catalyzed by CuCl in different ionic liquids and solvents has been investigated.

Application

3,5-Dimethyl-1-hexyn-3-ol may be used to synthesize:
  • 3,5-Dimethyl-1-phenyl-1-hexen-3-ol via one-pot palladium-mediated hydrostannylation/Stille cross-coupling.
  • 3,5-dimethyl-1-hexyn-3-acetate via esterification with acetic anhydride in a neutral ionic liquid (1-butyl-3-methylimidazolium tetrafluoroborate).
  • 3,5-Dimethyl-3-hydroxy-1-hexen-1-yl benzoate via anti-Markovnikov addition of benzoic acid.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A New approach for the generation and reaction of organotin hydrides: the development of reactions catalytic in tin.
Terstiege I and Maleczka R E
The Journal of Organic Chemistry, 64(2), 342-343 (1999)
Neutral ionic liquid [BMIm] BF4 promoted highly selective esterification of tertiary alcohols by acetic anhydride.
Duan Z, et al.
J. Mol. Catal. A: Chem., 246(1-2), 70-75 (2006)
Efficient Ruthenium?Catalysed Synthesis of 3?Hydroxy?1?propen?1?yl Benzoates: En Route to an Improved Isomerization of 2?Propyn?1?ols into α, β?Unsaturated Aldehydes.
Picquet M, et al.
European Journal of Organic Chemistry, 2000(13), 2361-2366 (2000)
The hydroxyl radical reaction rate constant and products of 3, 5-dimethyl-1-hexyn-3-ol.
Wells JR.
International Journal of Chemical Kinetics, 36(10), 534-544 (2004)
Yanlong Gu et al.
The Journal of organic chemistry, 69(2), 391-394 (2004-01-17)
Reactions of propargylic alcohols with CO(2) in a [BMIm][PhSO(3)]/CuCl catalytic system to produce the corresponding alpha-methylene cyclic carbonates were conducted with high yields. Mild reaction conditions, enhanced rates, improved yields, and recyclable ionic liquid catalyst systems are the remarkable features

Articles

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service