Skip to Content
Merck

250031

Sigma-Aldrich

(R)-(−)-N-(3,5-Dinitrobenzoyl)-α-phenylglycine

99%, for peptide synthesis

Synonym(s):

N-(3,5-Dinitrobenzoyl)-D-α-phenylglycine, R-(−)-N-(3,5-Dinitrobenzoyl)phenylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3CONHCH(C6H5)CO2H
CAS Number:
Molecular Weight:
345.26
Beilstein:
4719763
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Product Name

(R)-(−)-N-(3,5-Dinitrobenzoyl)-α-phenylglycine, 99%

Assay

99%

optical activity

[α]20/D −102°, c = 0.9 in THF

reaction suitability

reaction type: solution phase peptide synthesis

mp

216-217 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)[C@H](NC(=O)c1cc(cc(c1)[N+]([O-])=O)[N+]([O-])=O)c2ccccc2

InChI

1S/C15H11N3O7/c19-14(16-13(15(20)21)9-4-2-1-3-5-9)10-6-11(17(22)23)8-12(7-10)18(24)25/h1-8,13H,(H,16,19)(H,20,21)/t13-/m1/s1

InChI key

MIVUDAUOXJDARR-CYBMUJFWSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
P25485237647190357
optical activity

[α]20/D −102°, c = 0.9 in THF

optical activity

[α]20/D −155°, c = 1 in 1 M HCl

optical activity

[α]20/D +155°, c = 1 in 1 M HCl

optical activity

[α]24/D −31.7°, c = 0.76 in 1 M HCl

mp

216-217 °C (lit.)

mp

302 °C (dec.) (lit.)

mp

>300 °C (lit.)

mp

75-77 °C (lit.)

reaction suitability

reaction type: solution phase peptide synthesis

reaction suitability

reaction type: solution phase peptide synthesis

reaction suitability

reaction type: solution phase peptide synthesis

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

application(s)

detection

application(s)

peptide synthesis

application(s)

peptide synthesis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wolfgang Thormann et al.
Journal of pharmaceutical and biomedical analysis, 27(3-4), 555-567 (2002-01-05)
The electrokinetic separation and analysis of the enantiomers of albendazole sulfoxide (ABZSO), a sulfoxide with a sulfur stereogenic center hepatically formed during therapy with the anthelmintic drug albendazole (ABZ), is reported. Using aqueous or nonaqueous alkaline background electrolytes, ABZSO enantiomers
Resolution of enantiomers of 19-hydroxyeicosatetraenoate and 18-hydroxyeicosatetraenoate by chiral phase high-performance liquid chromatography of naphthoyl ester derivatives.
E H Oliw
Journal of chromatography, 526(2), 525-529 (1990-04-06)
K Roy et al.
Drug design and discovery, 17(4), 315-323 (2002-01-05)
QSAR analyses of matrix metalloproteinase (MMP) inhibitor N-[(substituted phenyl)sulfonyl]-N-4-nitrobenzylglycine hydroxamates, recently reported by Scozzafava and Supuran, have been attempted using linear free energy related (LFER) model of Hansch to explore the contribution patterns of the phenyl ring substitutions (P1' anchoring
S K Yang et al.
Journal of chromatography, 371, 195-209 (1986-12-26)
The direct resolution of K-region cis- and trans-dihydrodiol enantiomers of 14 unsubstituted and methyl- and bromo-substituted benz[a]anthracene (BA) derivatives was investigated by high-performance liquid chromatography with commercially available columns, packed with gamma-aminopropylsilanized silica to which either (R)-N-(3,5-dinitrobenzoyl)phenylglycine (R-DNBPG) or (S)-N-(3,5-dinitrobenzoyl)leucine
H B Weems et al.
Journal of chromatography, 371, 211-225 (1986-12-26)
The direct separation of 26 bay region and non-bay region mono-ol and diol enantiomers of phenanthrene, benz[a]anthracene, and chrysene was compared by high-performance liquid chromatography on commercially available columns, packed with gamma-aminopropylsilanized silica to which either (R)-N-(3,5-dinitrobenzoyl)phenylglycine(R-DNBPG) or (S)-N-(3,5-dinitrobenzoyl)leucine(S-DNBL) was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service