Skip to Content
Merck
All Photos(1)

Documents

245631

Sigma-Aldrich

Benzyl bromoacetate

96%

Synonym(s):

Bromoacetic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2COOCH2C6H5
CAS Number:
Molecular Weight:
229.07
Beilstein:
973658
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.544 (lit.)

bp

166-170 °C/22 mmHg (lit.)

density

1.446 g/mL at 25 °C (lit.)

SMILES string

BrCC(=O)OCc1ccccc1

InChI

1S/C9H9BrO2/c10-6-9(11)12-7-8-4-2-1-3-5-8/h1-5H,6-7H2

InChI key

JHVLLYQQQYIWKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzyl bromoacetate was used in the alkylation of (-)-2,3-O-isopropylidene-D-threitol that afforded lipopeptide, 2-[(4R,5R)-5-({[(9H-fluoren-9-yl)methoxy]carbonylaminomethyl}-2,2-dimethyl-1,3-dioxolan-4-yl)methoxy]acetic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toru Masaka et al.
Chemical & pharmaceutical bulletin, 61(11), 1184-1187 (2013-08-28)
A new component for the solid phase peptide synthesis of lipopeptide, 2-[(4R,5R)-5-({[(9H-fluoren-9-yl)methoxy]carbonylaminomethyl}-2,2-dimethyl-1,3-dioxolan-4-yl)methoxy]acetic acid (2), was designed and synthesized from (-)-2,3-O-isopropylidene-D-threitol (3) in 4 steps. The key step was the selective alkylation of 3 with benzyl bromoacetate in the presence of
Michael Kahnt et al.
Molecules (Basel, Switzerland), 24(12) (2019-06-20)
In this study, we report the synthesis of several amine-spacered conjugates of ursolic acid (UA) and 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetic acid (DOTA). Thus, a total of 11 UA-DOTA conjugates were prepared holding various oligo-methylene diamine spacers as well as different substituents at the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service