Skip to Content
Merck
All Photos(1)

Documents

227102

Sigma-Aldrich

2,2-Diethoxyacetophenone

>95%

Synonym(s):

α,α-Diethoxyacetophenone, Phenylglyoxal 2-diethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH(OCH2CH3)2
CAS Number:
Molecular Weight:
208.25
Beilstein:
2100306
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.01 mmHg ( 20 °C)

Assay

>95%

form

liquid

refractive index

n20/D 1.499 (lit.)

bp

131-134 °C/10 mmHg (lit.)

density

1.034 g/mL at 25 °C (lit.)

SMILES string

CCOC(OCC)C(=O)c1ccccc1

InChI

1S/C12H16O3/c1-3-14-12(15-4-2)11(13)10-8-6-5-7-9-10/h5-9,12H,3-4H2,1-2H3

InChI key

PIZHFBODNLEQBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Photocuring agent

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Wang et al.
ACS nano, 13(3), 3638-3648 (2019-03-12)
Construction of textured bioinspired surfaces with refined structures that exhibit superior wetting properties is of great importance for many applications ranging from self-cleaning, antibiofouling, anti-icing, oil/water separation, smart membrane, and microfluidic devices. Previously, the preparation of artificial surfaces generally relies
Yunlong Wang et al.
Journal of materials chemistry. B, 8(16), 3519-3526 (2020-01-29)
Point-of-care (POC) diagnosis is of great significance in offering precise and personalized treatment for patients with eye diseases. Contact lenses, as a kind of popular wearable device on the eye, provide a suitable platform for the integration of biosensors for
Yifei Wang et al.
Analytica chimica acta, 1070, 97-103 (2019-05-20)
As a typical antibiotic that is harmful to the health of human beings and widely used in animal husbandry, oxytetracycline (OTC) has a potential threat to the food safety. Therefore, the design of portable sensors for efficiently monitoring trace amounts
Isabel Ortiz de Solorzano et al.
Colloids and surfaces. B, Biointerfaces, 190, 110904-110904 (2020-03-22)
Photopolymerization is a selective technique that takes advantage of light-sensitive molecules to initiate and propagate monomeric structures to render covalently bonded macromolecular materials structures known as polymers. Herein, we present a novel one-step microfluidic synthesis of customized hybrid-thermoresponsive Poly(N-isopropylacrylamide) (PNIPAm)
Mohamed Elsherif et al.
ACS nano, 12(6), 5452-5462 (2018-05-12)
Low-cost, robust, and reusable continuous glucose monitoring systems that can provide quantitative measurements at point-of-care settings is an unmet medical need. Optical glucose sensors require complex and time-consuming fabrication processes, and their readouts are not practical for quantitative analyses. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service