Skip to Content
Merck
All Photos(1)

Key Documents

196533

Sigma-Aldrich

1,3-Benzenedimethanol

98%

Synonym(s):

m-Xylene-α,α′-diol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH2OH)2
CAS Number:
Molecular Weight:
138.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

154-159 °C/13 mmHg (lit.)

mp

56-60 °C (lit.)

SMILES string

OCc1cccc(CO)c1

InChI

1S/C8H10O2/c9-5-7-2-1-3-8(4-7)6-10/h1-4,9-10H,5-6H2

InChI key

YWMLORGQOFONNT-UHFFFAOYSA-N

Application

1,3-Benzenedimethanol was used in the synthesis of a series of cross-linked poly(orthocarbonate)s. It was also used in the synthesis of mixed-tethered system, required for the preparation of e-edge-[60]fullerenylmethanodihydropyrrole adduct.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cross-linked poly (orthocarbonate) s as organic solvent sorbents.
Sonmez HB and Wudl F.
Macromolecules, 38(5), 1623-1626 (2005)
Regioselective Synthesis of Novel e-Edge-[60] fullerenylmethanodihydropyrroles and 1, 2-Dihydromethano [60] fullerenes.
Chaker L, et al.
European Journal of Organic Chemistry, 2005(24), 5158-5162 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service