Skip to Content
Merck
All Photos(1)

Key Documents

141135

Sigma-Aldrich

2,3-Dicyanohydroquinone

98%

Synonym(s):

2,3-Dicyano-1,4-hydroquinone, 3,6-Dihydroxyphthalonitrile, DCH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NC)2C6H2-1,4-(OH)2
CAS Number:
Molecular Weight:
160.13
Beilstein:
2101249
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

>230 °C (dec.) (lit.)

functional group

nitrile

SMILES string

Oc1ccc(O)c(C#N)c1C#N

InChI

1S/C8H4N2O2/c9-3-5-6(4-10)8(12)2-1-7(5)11/h1-2,11-12H

InChI key

MPAIWVOBMLSHQA-UHFFFAOYSA-N

Application

2,3-Dicyanohydroquinone was used in the synthesis of phthalonitrile-3,6-ditriflate. It is a fluorescent dye and was used in determination of intracellular pH and calcium in avian neural crest cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Near-infrared absorbing unsymmetrical Zn (II) phthalocyanine for dye-sensitized solar cells.
Singh VK, et al.
Inorgorganica Chimica Acta, 407, 289-296 (2013)
C J Dickens et al.
The Journal of physiology, 428, 531-544 (1990-09-01)
1. Intracellular pH (pHi) and calcium (Cai2+) were studied in freely migrating neural crest cells and in closely packed non-migrating cells derived from avian neural tubes in vitro, using the fluorescent dyes 2,3-dicyanohydroquinone (DCH) and Indo-1 to measure pHi and
I Kurtz et al.
Biophysical journal, 48(3), 499-508 (1985-09-01)
We have developed new methodology for measuring intracellular pH (pHi) in cultured cell monolayers and epithelia by analyzing the emission spectra of the trapped fluorescent pH probe, 1,4-dihydroxyphthalonitrile (1,4-DHPN). This compound is unique since both its acid and base forms
J I Gillespie et al.
The Journal of physiology, 405, 385-395 (1988-11-01)
1. Measurements of intracellular pH (pHi) have been made using the fluorescent dye dicyano-hydroquinone (DCH) in somites isolated from 2-day-old chick embryos. Measurements of pHi, from freshly dissected somites, gave values of 7.18 +/- 0.02 (mean +/- S.D., n =
Fast intracellular pH determination in single cells by flow-cytometry.
G Valet et al.
Die Naturwissenschaften, 68(5), 265-266 (1981-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service