Skip to Content
Merck
All Photos(2)

Documents

106003

Sigma-Aldrich

2,3-Dibromopropene

80%, technical grade

Synonym(s):

(2-Bromo-2-propenyl) bromide, 2,3-Dibromopropylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2BrCBr=CH2
CAS Number:
Molecular Weight:
199.87
Beilstein:
878169
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

80%

refractive index

n20/D 1.544 (lit.)

bp

42-44 °C/17 mmHg (lit.)

density

2.045 g/mL at 25 °C

SMILES string

BrCC(Br)=C

InChI

1S/C3H4Br2/c1-3(5)2-4/h1-2H2

InChI key

YMFWYDYJHRGGPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Dibromopropene is an intermediate formed during the hydrolysis of nematocide:1,2-dibromo-3-chloropropane.

Application

2,3-Dibromopropene was used to test 2- and 3-carbon halogenated hydrocarbons for mutagenicity for Salmonella typhimurium strain TA 100 . It was used in the synthesis of N(Boc)-L-(2-Bromoallyl)-glycine.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

S J Stolzenberg et al.
Environmental mutagenesis, 2(1), 59-66 (1980-01-01)
Short-chain, 2- and 3- carbon halogenated hydrocarbons were tested for mutagenicity for Salmonella typhimurium strain TA 100 both with and without the presence of S-9. Without exception, all brominated derivatives were more mutagenic than the chlorinated derivatives, usually by a
Kinetics and products of hydrolysis of 1,2-dibromo-3-chloropropane.
N E Burlinson et al.
Environmental science & technology, 16(9), 627-632 (1982-09-01)
N-(Boc)-L-(2-Bromoallyl)-glycine: a versatile intermediate for the synthesis of optically active unnatural amino acids.
Leanna MR and Morton HE.
Tetrahedron Letters, 34(28), 4485-4488 (1993)
Gerald Pratsch et al.
The Journal of organic chemistry, 80(22), 11388-11397 (2015-10-31)
Visible-light photoreductive coupling of 2-arylallyl bromides in the presence of the photocatalyst Ru(bpy)3(PF6)2, a Hantzsch ester, and i-Pr2NEt gives 2,5-diaryl-1,5-dienes in high yield. This method avoids the use of stoichiometric metal reductants and is compatible with the presence of halogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service