LC/MS Analysis of Peptides on Discovery® BIO Wide Pore C18, Effect of TFA Concentration on MS Signal
![LC/MS Analysis of Peptides on Discovery® BIO Wide Pore C18, Effect of TFA Concentration on MS Signal application for HPLC](/deepweb/assets/sigmaaldrich/product/images/105/479/d62d2acb-2e1c-4500-9abc-5625992b9e00/800/d62d2acb-2e1c-4500-9abc-5625992b9e00.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery® BIO Wide Pore C18, 15 cm x 2.1 mm I.D., 3 μm particles (567202-U)
mobile phase
[A]: aqueous component (25 mM formic acid, 0.01% TFA or 0.1% TFA); [B] (50:50) water:acetonitrile containing same ionic additives as aq. component
flow rate
0.20 mL/min
column temp.
ambient
detector
(+)ESI
injection
1-3 μL
sample
RP Peptide Performance Standard (Alberta Peptide Inst., Cat. No. RPS-P0010)
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany