HPLC Analysis of RNA on Proteomix® WAX-NP5
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Proteomix WAX-NP5, 15 cm x 4.6 mm I.D., 5 μm particles (Z777262)
mobile phase
(A) 20 mmol Tris, pH 6.0; (B): A + 1.0 M NaCl
gradient
20%-100%B (40 min)
flow rate
0.6 mL/min
pressure
1015 psi (70 bar)
column temp.
Ambient
detector
UV, 260 nm
injection
5 μL
sample
RNA
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies