Merck
  • Identification of Δ6-monounsaturated fatty acids in human hair and nail samples by gas-chromatography-mass-spectrometry using ionic-liquid coated capillary column.
Identification of Δ6-monounsaturated fatty acids in human hair and nail samples by gas-chromatography-mass-spectrometry using ionic-liquid coated capillary column.
Journal of chromatography. A (2011-11-29)
Frédéric Destaillats, Marjorie Guitard, Cristina Cruz-Hernandez
ABSTRACT
Lipids found in human sebum contain specific fatty acids such as sapienic (cis-6 16:1), cis-8 18:1 and sebaleic (cis-5, cis-8 18:2) acids. These fatty acids belong to the n-10 series and the initial step involved in their synthesis is the desaturation of palmitic acid by the Δ6-desaturase to form sapienic acid. The occurrence in human hair and nail of sapienic (cis-6 16:1), cis-8 18:1 and sebaleic (cis-5, cis-8 18:2) acids has not been reported to our knowledge nor has the formation of Δ6-monounsaturated fatty acids from other saturated fatty acids such as stearic acid. The pre-requisite for such identification is the ability to separate cis-6 from cis-8 monounsaturated fatty acid derivative (i.e. cis-6 18:1 from cis-8 18:1 methyl esters) by gas-chromatography (GC) and such separation is not achievable using cyanoalkyl based highly polar capillary columns. In the present study, we used the 100 m SLB-IL 111 ionic liquid based capillary column recently commercialized by Supelco (Bellefonte, PA). The identification was performed by gas-chromatography-mass-spectrometry (GC-MS) with electronic impact (EI) ionization using 4,4-dimethyloxazoline (DMOX) derivatives. Baseline separation between critical cis-6 18:1 and cis-8 18:1 isomers was obtained allowing unambiguous identification based on MS fragmentation and pure standards. In sebum, hair and nail samples, sapienic, cis-8 18:1 and sebaleic acids were found and more importantly, petroselinic acid was identified in these human tissues for the first time. In addition, we identified in hair and nail lipids cis-6 14:1, cis-6 15:1, iso-cis-6 16:1, aiso-cis-6 17:1 and cis-6 17:1 as their DMOX derivatives based on molecular ion as well as diagnostic ion fragments at m/z 167, 180 and 194. Possible biosynthesis scenario is postulated to explain the occurrence of these Δ6-monounsaturated fatty acids in human sebum, hair and nail lipids.
Product Description
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.25 μm
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.10 μm
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 15 m × 0.32 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.10 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 10 m × 0.10 mm, df 0.10 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 15 m × 0.10 mm, df 0.10 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.50 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 15 m × 0.25 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 20 m × 0.18 mm, df 0.18 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 1.00 μm