Merck
  • Transgenic expression of HuR increases vasogenic edema and impedes functional recovery in rodent ischemic stroke.
Transgenic expression of HuR increases vasogenic edema and impedes functional recovery in rodent ischemic stroke.
Neuroscience letters (2017-10-07)
Agnieszka A Ardelt, Randall S Carpenter, Ifeanyi Iwuchukwu, An Zhang, William Lin, Ewa Kosciuczuk, Cyrus Hinkson, Tania Rebeiz, Sydney Reitz, Peter H King
ABSTRACT
Ischemic stroke produces significant morbidity and mortality, and acute interventions are limited by short therapeutic windows. Novel approaches to neuroprotection and neurorepair are necessary. HuR is an RNA-binding protein (RBP) which modulates RNA stability and translational efficiency of genes linked to ischemic stroke injury. Using a transgenic (Tg) mouse model, we examined the impact of ectopic HuR expression in astrocytes on acute injury evolution after transient middle cerebral artery occlusion (tMCAO). HuR transgene expression was detected in astrocytes in perilesional regions and contralaterally. HuR Tg mice did not improve neurologically 72h after injury, whereas littermate controls did. In Tg mice, increased cerebral vascular permeability and edema were observed. Infarct volume was not affected by the presence of the transgene. Ectopic expression of HuR in astrocytes worsens outcome after transient ischemic stroke in mice in part by increasing vasogenic cerebral edema. These findings suggest that HuR could be a therapeutic target in cerebral ischemia/reperfusion.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
Donkey Anti-Chicken IgY Antibody, Cy3 conjugate, Species Adsorbed, Chemicon®, from donkey
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution