MilliporeSigma
  • Cucurbitacin B Protects Against Pressure Overload Induced Cardiac Hypertrophy.
Cucurbitacin B Protects Against Pressure Overload Induced Cardiac Hypertrophy.
Journal of cellular biochemistry (2017-04-09)
Yang Xiao, Zheng Yang, Qing-Qing Wu, Xiao-Han Jiang, Yuan Yuan, Wei Chang, Zhou Yan Bian, Jin Xiu Zhu, Qi-Zhu Tang
ABSTRACT
Lack of effective anti-cardiac hypertrophy drugs creates a major cause for the increasing prevalence of heart failure. In the present study, we determined the anti-hypertrophy and anti-fibrosis potential of a natural plant triterpenoid, Cucurbitacin B both in vitro and in vivo. Aortic banding (AB) was performed to induce cardiac hypertrophy. After 1 week of surgery, mice were receive cucurbitacin B treatment (Gavage, 0.2 mg/kg body weight/2 day). After 4 weeks of AB, cucurbitacin B demonstrated a strong anti-hypertrophy and -fibrosis ability as evidenced by decreased of heart weight, myocardial cell cross-sectional area and interstitial fibrosis, ameliorated of systolic and diastolic abnormalities, normalized in gene expression of hypertrophic and fibrotic markers, reserved microvascular density in pressure overload induced hypertrophic mice. Cucurbitacin B also showed significant hypertrophy inhibitory effect in phenylephrine stimulated cardiomyocytes. The Cucurbitacin B-mediated mitigated cardiac hypertrophy was attributable to the increasing level of autophagy, which was associated with the blockade of Akt/mTOR/FoxO3a signal pathway, validated by SC79, MK2206, and 3-MA, the Akt agonist, inhibitor and autophagy inhibitor in vitro. The overexpression of constitutively active Akt completely abolished the Cucurbitacin B-mediated protection of cardiac hypertrophy in human cardiomyocytes AC16. Collectively, our findings suggest that cucurbitacin B protects against cardiac hypertrophy through increasing the autophagy level in cardiomyocytes, which is associated with the inhibition of Akt/mTOR/FoxO3a signal axis. J. Cell. Biochem. 118: 3899-3910, 2017. © 2017 Wiley Periodicals, Inc.
Sigma-Aldrich
Anti-α-Actinin Antibody, clone AT6/172, clone AT6/172, Upstate®, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
SC79, ≥97% (NMR)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Phenylephrine Related Compound C, Pharmaceutical Secondary Standard; Certified Reference Material