Skip to Content
AD
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
The role of bicarbonate anions in methotrexate degradation via UV/TiO
The role of bicarbonate anions in methotrexate degradation via UV/TiO
Water research (2017-02-06)
Webber Wei-Po Lai, Ming-Hao Hsu, Angela Yu-Chen Lin
PMID
28157604
ABSTRACT
Bicarbonate anion (HCO
Product Description
284572
Sigma-Aldrich
4-Aminobenzamide, 98%
Pricing
A0879
Sigma-Aldrich
N-(4-Aminobenzoyl)-L-glutamic acid, ≥98% (TLC)
Pricing
861634
Sigma-Aldrich
2,4-Diamino-6-(hydroxymethyl)pteridine, 95%
Pricing