Merck
  • Clinicoradiological characteristics, management and prognosis of primary myeloid sarcoma of the central nervous system: A report of four cases.
Clinicoradiological characteristics, management and prognosis of primary myeloid sarcoma of the central nervous system: A report of four cases.
Oncology letters (2017-09-21)
Bao Yang, Chenlong Yang, Jingyi Fang, Jun Yang, Yulun Xu
ABSTRACT
Myeloid sarcoma (MS) is a localized tumor composed of premature precursors of granulocytic cells, which may occur in any organ and most commonly involves the soft tissue and musculoskeletal system. This malignancy may occur in the presence or absence of hematological disorders. Primary MS involving the central nervous system (CNS-MS) is rare, and has only been described in a small number of isolated case reports. The diagnosis of CNS-MS is challenging and strategies for its management are undefined. The present study describes 4 cases of CNS-MS. The hematological indices at admission were normal and all patients presented with location-associated nonspecific symptoms. In magnetic resonance imaging scans, the tumors appeared isointense on T1-weighted and T2-weighted images, with marked enhancement following contrast agent administration. Only 1 patient progressed to acute myeloblastic leukemia (AML), with this occurring 3 weeks following histological diagnosis. During the follow-up period, the patient with AML succumbed to the disease, local recurrence was noted in another patient for which a second surgery was requested and no progression was observed in the remaining 2 patients. CNS-MS has unique radiological characteristics and, due to its diffuse tissue infiltration, gross total resection is challenging. It is important for clinicians to be aware of potential hematological disorders in patients with CNS-MS. A combined surgical and chemotherapeutic strategy may be able to provide long-term control of this malignancy.
Sigma-Aldrich
Anti-MPO antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Sigma-Aldrich
Anti-LYG2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CD3 antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP162, affinity isolated antibody
Sigma-Aldrich
Anti-CD38 antibody, Rabbit monoclonal, clone SP149, recombinant, expressed in proprietary host, tissue culture supernatant
Sigma-Aldrich
Monoclonal Anti-CD117 antibody produced in mouse, clone 104D2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-MUC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CD99 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CD33 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CD34 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CD20 antibody, Rabbit monoclonal, clone SP32, recombinant, expressed in proprietary host, affinity isolated antibody
Sigma-Aldrich
Anti-KI-67 antibody, Rabbit monoclonal, clone SP6, recombinant, expressed in proprietary host, tissue culture supernatant
Sigma-Aldrich
Anti-GFAP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-S-100 (β-Subunit) antibody, Mouse monoclonal, clone SH-B1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NCAM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution