Merck
  • Pharmacokinetic study of calenduloside E and its active metabolite oleanolic acid in beagle dog using liquid chromatography-tandem mass spectrometry.
Pharmacokinetic study of calenduloside E and its active metabolite oleanolic acid in beagle dog using liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-02-22)
Meiyun Shi, Yan Yang, Yantong Sun, Longmei Cheng, Sen Zhao, Huibo Xu, J Paul Fawcett, Xiaobo Sun, Jingkai Gu
ABSTRACT
Aralia mandshrica is a well-known traditional Chinese medicine from Northeast China commonly used to treat digestive, circulatory and immune system disorders. Calenduloside E is one of its bioactive components currently under evaluation as a pure drug. In this study, a highly sensitive and rapid method based on liquid chromatography-tandem mass spectrometry (LC-MS/MS) for the simultaneous quantitation of calenduloside E and its active metabolite oleanolic acid in beagle dog plasma has been developed and validated. Samples containing the ammonium salt of simvastatin acid as internal standard (IS) were purified by solid phase extraction and separated on a SUPELCO Ascentis-C18 column (50mm×4.6mm i.d., 5μm) using gradient elution with 0.35% formic acid and acetonitrile. Analytes and IS were detected in a cycle time of 5min after ionization in the negative ion mode by multiple reaction monitoring of the precursor-to-product ion transitions at m/z 631.4→455.4 and m/z 435.4→319.0 for calenduloside E and IS respectively and by single ion monitoring of the ion at m/z 455.4 for oleanolic acid. The method was linear over the concentration range 0.4-100ng/mL for both analytes using 0.5mL plasma. Inter- and intra-day precisions were both <6.96% with accuracies <6.40%. In the pharmacokinetic (PK) study, beagle dogs were given oral doses of calenduloside E (1.05, 2.10 and 4.20mg/kg) and an intravenous injection of 2.10mg/kg. The absolute bioavailability of calenduloside E was only 0.58%. Area under the plasma concentration time curve (AUC(0-t)) for the oral doses of calenduloside E was approximately dose proportional while other PK parameters (t1/2, Tmax and MRT) showed no significant differences among the three doses (P>0.05). The PK data provide a useful platform on which to base future clinical studies of calenduloside E.
Product Description
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 3 mm
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® ES-Cyano HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 10 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 21.2 mm
Supelco
Ascentis® ES-Cyano HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Ascentis® ES-Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm