Merck
  • The PYRIN domain-only protein POP2 inhibits inflammasome priming and activation.
The PYRIN domain-only protein POP2 inhibits inflammasome priming and activation.
Nature communications (2017-06-06)
Rojo A Ratsimandresy, Lan H Chu, Sonal Khare, Lucia de Almeida, Anu Gangopadhyay, Mohanalaxmi Indramohan, Alexander V Misharin, David R Greaves, Harris Perlman, Andrea Dorfleutner, Christian Stehlik
ABSTRACT
Inflammasomes are protein platforms linking recognition of microbe, pathogen-associated and damage-associated molecular patterns by cytosolic sensory proteins to caspase-1 activation. Caspase-1 promotes pyroptotic cell death and the maturation and secretion of interleukin (IL)-1β and IL-18, which trigger inflammatory responses to clear infections and initiate wound-healing; however, excessive responses cause inflammatory disease. Inflammasome assembly requires the PYRIN domain (PYD)-containing adaptor ASC, and depends on PYD-PYD interactions. Here we show that the PYD-only protein POP2 inhibits inflammasome assembly by binding to ASC and interfering with the recruitment of ASC to upstream sensors, which prevents caspase-1 activation and cytokine release. POP2 also impairs macrophage priming by inhibiting the activation of non-canonical IκB kinase ɛ and IκBα, and consequently protects from excessive inflammation and acute shock in vivo. Our findings advance our understanding of the complex regulatory mechanisms that maintain a balanced inflammatory response and highlight important differences between individual POP members.
Sigma-Aldrich
Interleukin-1β Protein, Recombinant human, The recombinant human Interleukin-1beta protein (or IL-1beta protein) is a potent immuno-modulator which mediates a wide range of immune & inflammatory responses including the activation of B & T-cells.
Sigma-Aldrich
IFN- γ Protein, Human Recombinant Animal Free, The Interferon gamma (or IFN-γ protein) is an acid-labile interferon produced by CD4 & CD8 T lymphocytes as well as activated NK cells. Manufactured using all non-animal reagents.
Sigma-Aldrich
Transforming Growth Factor-β2 Protein, Recombinant human, Transforming Growth Factor-beta2 (TGF-beta2) is a member of a superfamily of homologous, disulfide-linked, homodimeric proteins that regulate the proliferation & differentiation of normal & transformed cells.
Sigma-Aldrich
Interferon-β Protein, Recombinant human, Recombinant Human Interferon Beta 1a (Hu-IFNbeta 1a).
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, BioXtra, ≥99% (HPLC), from microbial
Sigma-Aldrich
BAY 11-7082, The BAY 11-7082, also referenced under CAS 19542-67-7, controls the biological activity of TNF-α. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Tumor Necrosis Factor-α Protein, Recombinant human, Tumor Necrosis Factor-alpha (TNF-alpha) is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells & certain other target cells.