Merck
  • New anthracene-based Schiff bases: Theoretical and experimental investigations of photophysical and electrochemical properties.
New anthracene-based Schiff bases: Theoretical and experimental investigations of photophysical and electrochemical properties.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2016-12-25)
Danuta Sek, Mariola Siwy, Marzena Grucela, Grzegorz Małecki, Elżbieta M Nowak, Gabriela Lewinska, Jerzy Santera, Katarzyna Laba, Mieczyslaw Lapkowski, Sonia Kotowicz, Ewa Schab-Balcerzak
ABSTRACT
The new Schiff bases bearing anthracene unit were synthesized from 2-aminoanthracene and various aldehydes such as: benzaldehyde, 4-(diphenylamino)benzaldehyde, 9-phenanthrenecarboxaldehyde, 9-anthracenecarboxaldehyde, and biphenyl-4-carboxaldehyde, 2-naphthaldehyde. Resulted azomethines were characterized by IR, NMR (
Sigma-Aldrich
2-Aminoanthracene, 96%
Sigma-Aldrich
9-Phenanthrenecarboxaldehyde, 97%
Sigma-Aldrich
2-Naphthaldehyde, 98%
Sigma-Aldrich
Biphenyl-4-carboxaldehyde, 99%
Sigma-Aldrich
9-Anthracenecarboxaldehyde, 97%
Sigma-Aldrich
4-(Diphenylamino)benzaldehyde, 97%