Merck
Synthesis of ketones from biomass-derived feedstock.
Nature communications (2017-02-01)
Qinglei Meng, Minqiang Hou, Huizhen Liu, Jinliang Song, Buxing Han
ABSTRACT
Cyclohexanone and its derivatives are very important chemicals, which are currently produced mainly by oxidation of cyclohexane or alkylcyclohexane, hydrogenation of phenols, and alkylation of cyclohexanone. Here we report that bromide salt-modified Pd/C in H
Sigma-Aldrich
2-Methoxycyclohexanone, 97%
Sigma-Aldrich
Ethylcyclohexane, ≥99%
Sigma-Aldrich
p-Propyl anisole, ≥99%, FCC, FG
Sigma-Aldrich
Methyl 2-methoxybenzoate, ≥97%
Sigma-Aldrich
2-Cyclohexylethanol, 99%
Sigma-Aldrich
4-Ethylcyclohexanone, 97%
Sigma-Aldrich
4-Methoxybenzamide, 98%
Sigma-Aldrich
4-(Trifluoromethyl)cyclohexanone, 97%