Sigma-Aldrich
Poly(ethylene glycol) methyl ether-block-poly(lactide-co-glycolide), PEG average Mn 5,000, PLGA Mn 20,000, lactide:glycolide 50:50
Sigma-Aldrich
Poly(ethylene glycol) methyl ether-block-poly(lactide-co-glycolide), PEG average Mn 5,000, PLGA Mn 15,000, lactide:glycolide 50:50
Sigma-Aldrich
Poly(ethylene glycol) methyl ether-block-poly(lactide-co-glycolide), PEG average Mn 5,000, PLGA average Mn 5,000, lactide:glycolide 80:20
Sigma-Aldrich
4arm-poly(lactide-co-glycolide), average Mn 38,000-60,000, lactide:glycolide 55:45
Sigma-Aldrich
N-Hydroxysuccinimide ester-poly(ethylene glycol)-b-poly(D,L lactide), PEG average Mn 5,000, PDLA average Mn 16,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether-block-poly(lactide-co-glycolide), PEG average Mn 2,000, PLGA average Mn 10,000, lactide:glycolide 80:20
Sigma-Aldrich
Poly(ethylene glycol) methyl ether-block-poly(lactide-co-glycolide), PEG average Mn 5,000, PLGA Mn 15,000, lactide:glycolide 80:20
Sigma-Aldrich
Poly(D,L-lactide-b-glycolide) lactide:glycolide 50:50, diamine, viscosity 0.035 dL/g
Sigma-Aldrich
Poly(ethylene glycol) methyl ether-block-poly(lactide-co-glycolide), PEG average Mn 5,000, PLGA average Mn 10,000, lactide:glycolide 80:20
Sigma-Aldrich
Poly(ethylene glycol) methyl ether-block-poly(lactide-co-glycolide), PEG average Mn 5,000, PLGA Mn 5,000, lactide:glycolide 50:50
Sigma-Aldrich
Poly(ethylene glycol) methyl ether-block-poly(lactide-co-glycolide), PEG average Mn 5,000, PLGA Mn 10,000, lactide:glycolide 50:50
Sigma-Aldrich
Poly(ethylene glycol) methyl ether-block-poly(lactide-co-glycolide), PEG average Mn 2,000, PLGA Mn 3,000, lactide:glycolide 50:50
Sigma-Aldrich
Poly(D,L-lactide-b-glycolide) lactide:glycolide 50:50, amine terminated, average Mn 5,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether-block-poly(D,L lactide), PEG average Mn 5000, PDLLA average Mn 20000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether-block-poly(D,L lactide), PEG average Mn 5000, PDLA average Mn 50000
Sigma-Aldrich
Poly(lactide-co-glycolide)-Flamma Fluor near-IR, lactide:glycolide (50:50), Mn 20,000-30,000
Sigma-Aldrich
Poly(lactide-co-glycolide)-fluorescein, lactide:glycolide 50:50, Mn 10,000-20,000
Sigma-Aldrich
Poly(lactide-co-glycolide)-Rhodamine B, lactide:glycolide 50:50, Mn 10,000-30,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide)(50:50)-b-poly(ethylene glycol), 10k-2k
Sigma-Aldrich
Carboxylic acid-poly(ethylene glycol)-b-poly(D,L lactide), PEG average Mn 5,000, PDLA average Mn 55,000
Sigma-Aldrich
Carboxylic acid-poly(ethylene glycol)-b-poly(D,L lactide), PEG average Mn 5,000, PDLA average Mn 16,000
Sigma-Aldrich
Poly(D,L-lactide-b-glycolide) lactide:glycolide 75:25, amine (dihydrazide) terminated, average Mn 12,000
Sigma-Aldrich
3arm-poly(lactide-co-glycolide), average Mn 38,000-60,000, lactide:glycolide 55:45