Merck
  • WP1066, a novel JAK2 inhibitor, suppresses proliferation and induces apoptosis in erythroid human cells carrying the JAK2 V617F mutation.
WP1066, a novel JAK2 inhibitor, suppresses proliferation and induces apoptosis in erythroid human cells carrying the JAK2 V617F mutation.
Clinical cancer research : an official journal of the American Association for Cancer Research (2008-02-05)
Srdan Verstovsek, Taghi Manshouri, Alfonso Quintás-Cardama, David Harris, Jorge Cortes, Francis J Giles, Hagop Kantarjian, Waldemar Priebe, Zeev Estrov
ABSTRACT
The discovery of an activating somatic mutation in codon 617 of the gene encoding the Janus kinase (JAK)-2 (JAK2 V617F) in patients with myeloproliferative disorders has opened new avenues for the development of targeted therapies for these malignancies. However, no effective JAK2 inhibitors are currently available for clinical use. We investigated the activity of (E)-3(6-bromopyridin-2-yl)-2-cyano-N-(S0-1phenylethyl)acrylamide (WP1066), a novel analogue of the JAK2 inhibitor AG490, in JAK2 V617F-positive erythroleukemia HEL cells and in blood cells from patients with polycythemia vera. We found that WP1066 significantly inhibited JAK2 and its downstream signal transducer and activator of transcription-3, signal transducer and activator of transcription-5, and extracellular signal-regulated kinase-1/2 pathways in a dose- and time-dependent manner. As a result, WP1066 concentrations in the low micromolar range induced time- and dose-dependent antiproliferative and proapoptotic effects in HEL cells. As expected, WP1066 inhibited the proliferation of peripheral blood hematopoietic progenitors of patients with polycythemia vera carrying the JAK2 V617F mutation in a dose-dependent manner. Our data suggest that WP1066 is active both in vitro and ex vivo and should be further developed for the treatment of neoplasms expressing the JAK2 V617F mutation.
Product Description
Supelco
Discovery® Cyano Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm