Merck
  • Autophagy is induced in the skeletal muscle of cachectic cancer patients.
Autophagy is induced in the skeletal muscle of cachectic cancer patients.
Scientific reports (2016-07-28)
Zaira Aversa, Fabrizio Pin, Simone Lucia, Fabio Penna, Roberto Verzaro, Maurizio Fazi, Giuseppina Colasante, Andrea Tirone, Filippo Rossi Fanelli, Cesarina Ramaccini, Paola Costelli, Maurizio Muscaritoli
ABSTRACT
Basal rates of autophagy can be markedly accelerated by environmental stresses. Recently, autophagy has been involved in cancer-induced muscle wasting. Aim of this study has been to evaluate if autophagy is induced in the skeletal muscle of cancer patients. The expression (mRNA and protein) of autophagic markers has been evaluated in intraoperative muscle biopsies. Beclin-1 protein levels were increased in cachectic cancer patients, suggesting autophagy induction. LC3B-I protein levels were not significantly modified. LC3B-II protein levels were significantly increased in cachectic cancer patients suggesting either increased autophagosome formation or reduced autophagosome turnover. Conversely, p62 protein levels were increased in cachectic and non-cachectic cancer patients, suggesting impaired autophagosome clearance. As for mitophagy, both Bnip3 and Nix/Bnip3L show a trend to increase in cachectic patients. In the same patients, Parkin levels significantly increased, while PINK1 was unchanged. At gene level, Beclin-1, p-62, BNIP3, NIX/BNIP3L and TFEB mRNAs were not significantly modulated, while LC3B and PINK1 mRNA levels were increased and decreased, respectively, in cachectic cancer patients. Autophagy is induced in the skeletal muscle of cachectic cancer patients, although autophagosome clearance appears to be impaired. Further studies should evaluate whether modulation of autophagy could represent a relevant therapeutic strategy in cancer cachexia.
Sigma-Aldrich
Anti-p62/SQSTM1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-PINK1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-Parkin antibody, Mouse monoclonal, clone PRK8, purified from hybridoma cell culture