Merck
  • Chemical genetics reveals a complex functional ground state of neural stem cells.
Chemical genetics reveals a complex functional ground state of neural stem cells.
Nature chemical biology (2007-04-10)
Phedias Diamandis, Jan Wildenhain, Ian D Clarke, Adrian G Sacher, Jeremy Graham, David S Bellows, Erick K M Ling, Ryan J Ward, Leanne G Jamieson, Mike Tyers, Peter B Dirks
ABSTRACT
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the proliferation and self-renewal of NSCs, which we refer to as the 'ground state', remains largely uncharacterized. Although the candidate gene approach has uncovered vital pathways in NSC biology, so far only a few highly studied pathways have been investigated. Based on the intimate relationship between NSC self-renewal and neurosphere proliferation, we undertook a chemical genetic screen for inhibitors of neurosphere proliferation in order to probe the operational circuitry of the NSC. The screen recovered small molecules known to affect neurotransmission pathways previously thought to operate primarily in the mature central nervous system; these compounds also had potent inhibitory effects on cultures enriched for brain cancer stem cells. These results suggest that clinically approved neuromodulators may remodel the mature central nervous system and find application in the treatment of brain cancer.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
LOPAC®1280 - Small Scale, International Version
Supelco
Androsterone, VETRANAL®, analytical standard
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Supelco
Tulobuterol hydrochloride, VETRANAL®, analytical standard
Supelco
Ibuprofen
Sigma-Aldrich
4-Aminopyridine, ≥99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
D-erythro-Dihydrosphingosine, ≥98%
Sigma-Aldrich
4-Aminopyridine, 98%
Supelco
Semicarbazide hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
(Z)-Guggulsterone, ≥89% (HPLC), powder
Sigma-Aldrich
1-Methylimidazole, ≥99%, purified by redistillation
Sigma-Aldrich
(−)-p-Bromolevamisole oxalate, 99%
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Acetamide, ≥99.0% (GC)
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Supelco
4-Androstene-3,17-dione, VETRANAL®, analytical standard
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, for complexometry, ≥99.0%
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Tryptamine hydrochloride, 99%
Sigma-Aldrich
4-Imidazoleacetic acid hydrochloride, 98%
Sigma-Aldrich
1-Methylimidazole, ReagentPlus®, 99%
Sigma-Aldrich
Harmane, 98%