Merck
  • Genome-wide association study identifies 3 genomic loci significantly associated with serum levels of homoarginine: the AtheroRemo Consortium.
Genome-wide association study identifies 3 genomic loci significantly associated with serum levels of homoarginine: the AtheroRemo Consortium.
Circulation. Cardiovascular genetics (2013-09-21)
Marcus E Kleber, Ilkka Seppälä, Stefan Pilz, Michael M Hoffmann, Andreas Tomaschitz, Niku Oksala, Emma Raitoharju, Leo-Pekka Lyytikäinen, Kari-Matti Mäkelä, Reijo Laaksonen, Mika Kähönen, Olli T Raitakari, Jie Huang, Katharina Kienreich, Astrid Fahrleitner-Pammer, Christiane Drechsler, Vera Krane, Bernhard O Boehm, Wolfgang Koenig, Christoph Wanner, Terho Lehtimäki, Winfried März, Andreas Meinitzer
ABSTRACT
Low serum levels of the amino acid derivative, homoarginine, have been associated with increased risk of total and cardiovascular mortality. Homoarginine deficiency may be related to renal and heart diseases, but the pathophysiologic role of homoarginine and the genetic regulation of its serum levels are largely unknown. In 3041 patients of the Ludwigshafen Risk and Cardiovascular Health (LURIC) study referred for coronary angiography and 2102 participants of the Young Finns Study (YFS), we performed a genome-wide association study to identify genomic loci associated with homoarginine serum levels and tested for associations of identified single-nucleotide polymorphisms with mortality in LURIC. We found genome-wide significant associations with homoarginine serum levels on chromosome 2 at the carbamoyl phosphate synthetase I locus, on chromosome 5 at the alanine-glyoxylate aminotransferase 2 locus, and on chromosome 15 at the glycine amidinotransferase locus, as well as a suggestive association on chromosome 6 at the Homo sapiens mediator complex subunit 23 gene/arginase I locus. All loci harbor enzymes located in the mitochondrium are involved in arginine metabolism. The strongest association was observed for rs1153858 at the glycine amidinotransferase locus with a P value of 1.25E-45 in the combined analysis and has been replicated in both the Die Deutsche Diabetes Dialyse Studie (4D study) and the Graz Endocrine Causes of Hypertension (GECOH) study. In our genome-wide association study, we identified 3 chromosomal regions significantly associated with serum homoarginine and another region with suggestive association, providing novel insights into the genetic regulation of homoarginine.
Sigma-Aldrich
Anti-GATM antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.