Merck
  • miR-10a regulates proliferation of human cardiomyocyte progenitor cells by targeting GATA6.
miR-10a regulates proliferation of human cardiomyocyte progenitor cells by targeting GATA6.
PloS one (2014-07-30)
Dandan Liang, Lixiao Zhen, Tianyou Yuan, Jian Huang, Fangfei Deng, Wuyahan, Hong Zhang, Lei Pan, Yi Liu, Erlinda The, Zuoren Yu, Weidong Zhu, Yuzhen Zhang, Li Li, Luying Peng, Jun Li, Yi-Han Chen
ABSTRACT
microRNAs (miRNAs) play essential roles in cardiogenesis. The altered expression of miRNAs can result in cardiac malformations by inducing abnormalities in the behavior of cardiac cells. However, the role of miR-10a in the regulation of cardiomyocyte progenitor cells (CMPCs) remains undetermined. In the present study, we found that up- or down-regulation of miR-10a inhibited or promoted the proliferation of human CMPCs, respectively, without affecting their differentiation toward cardiomyocytes. miR-10a bound to GATA6 directly and reduced GATA6 expression. Over-expression of GATA6 greatly attenuated the miR-10a-mediated inhibitory effect on the proliferation of human CMPCs. Thus, our results indicate that miR-10a could effectively modulate the proliferation of human CMPCs by targeting GATA6. The finding provides novel insights into the potency of miR-10a during heart development.
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Roche
Cell Proliferation ELISA, BrdU (colorimetric), sufficient for ≤1,000 tests