Merck
  • Abnormal cartilage development and altered N-glycosylation in Tmem165-deficient zebrafish mirrors the phenotypes associated with TMEM165-CDG.
Abnormal cartilage development and altered N-glycosylation in Tmem165-deficient zebrafish mirrors the phenotypes associated with TMEM165-CDG.
Glycobiology (2015-01-23)
Riet Bammens, Nickita Mehta, Valérie Race, François Foulquier, Jaak Jaeken, Michael Tiemeyer, Richard Steet, Gert Matthijs, Heather Flanagan-Steet
ABSTRACT
The congenital disorders of glycosylation (CDG), a group of inherited diseases characterized by aberrant glycosylation, encompass a wide range of defects, including glycosyltransferases, glycosidases, nucleotide-sugar transporters as well as proteins involved in maintaining Golgi architecture, pH and vesicular trafficking. Mutations in a previously undescribed protein, TMEM165, were recently shown to cause a new form of CDG, termed TMEM165-CDG. TMEM165-CDG patients exhibit cartilage and bone dysplasia and altered glycosylation of serum glycoproteins. We utilized a morpholino knockdown strategy in zebrafish to investigate the physiologic and pathogenic functions of TMEM165. Inhibition of tmem165 expression in developing zebrafish embryos caused craniofacial abnormalities, largely attributable to fewer chondrocytes. Decreased expression of several markers of cartilage and bone development suggests that Tmem165 deficiency alters both chondrocyte and osteoblast differentiation. Glycomic analysis of tmem165 morphants also revealed altered initiation, processing and extension of N-glycans, paralleling some of the glycosylation changes noted in human patients. Collectively, these findings highlight the utility of zebrafish to elucidate pathogenic mechanisms associated with glycosylation disorders and suggest that the cartilage and bone dysplasia manifested in TMEM165-CDG patients may stem from abnormal development of chondrocytes and osteoblasts.
Sigma-Aldrich
Iodomethane-12C, ≥99.9 atom % 12C, ≥99% (CP), contains copper as stabilizer
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Acridine Orange hydrochloride solution, ≥95.0% (HPLC), 10 mg/mL in H2O
Sigma-Aldrich
Iodomethane, purum, ≥99.0% (GC)
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99.5%
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99%
Sigma-Aldrich
Monoclonal Anti-Chondroitin Sulfate antibody produced in mouse, clone CS-56, ascites fluid
Sigma-Aldrich
Iodomethane solution, 2.0 M in tert-butyl methyl ether, contains copper as stabilizer
Sigma-Aldrich
Maltotriose, ≥90% (HPLC)