Merck
  • Parameterization of hyperpolarized (13)C-bicarbonate-dissolution dynamic nuclear polarization.
Parameterization of hyperpolarized (13)C-bicarbonate-dissolution dynamic nuclear polarization.
Magma (New York, N.Y.) (2015-10-10)
David Johannes Scholz, Angela M Otto, Josef Hintermair, Franz Schilling, Annette Frank, Ulrich Köllisch, Martin A Janich, Rolf F Schulte, Markus Schwaiger, Axel Haase, Marion I Menzel
ABSTRACT
(13)C metabolic MRI using hyperpolarized (13)C-bicarbonate enables preclinical detection of pH. To improve signal-to-noise ratio, experimental procedures were refined, and the influence of pH, buffer capacity, temperature, and field strength were investigated. Bicarbonate preparation was investigated. Bicarbonate was prepared and applied in spectroscopy at 1, 3, 14 T using pure dissolution, culture medium, and MCF-7 cell spheroids. Healthy rats were imaged by spectral-spatial spiral acquisition for spatial and temporal bicarbonate distribution, pH mapping, and signal decay analysis. An optimized preparation technique for maximum solubility of 6 mol/L and polarization levels of 19-21% is presented; T1 and SNR dependency on field strength, buffer capacity, and pH was investigated. pH mapping in vivo is demonstrated. An optimized bicarbonate preparation and experimental procedure provided improved T1 and SNR values, allowing in vitro and in vivo applications.
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Cesium hydroxide monohydrate, ≥99.5% trace metals basis
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Cesium hydroxide monohydrate, 99.95% trace metals basis
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Cesium hydroxide hydrate, ≥90%, ≥99.5% (metals basis)