Merck
  • Preparation, characterization and in vitro dissolution behavior of porous biphasic α/β-tricalcium phosphate bioceramics.
Preparation, characterization and in vitro dissolution behavior of porous biphasic α/β-tricalcium phosphate bioceramics.
Materials science & engineering. C, Materials for biological applications (2015-12-15)
Lu Xie, Haiyang Yu, Yi Deng, Weizhong Yang, Li Liao, Qin Long
ABSTRACT
The ideal bone tissue engineering scaffolds are long-cherished with the properties of interconnected macroporous structures, adjustable degradation and excellent biocompatibility. Here, a series of porous α/β-tricalcium phosphate (α/β-TCP) biphasic bioceramics with different phase ratios of α-TCP and β-TCP were successfully synthesized by heating an amorphous calcium phosphate precursor. The chemical and morphological characterization showed that α- and β-TCP phases co-existed in the α/β-TCP bioceramics and they had interconnected pore structures with size between 200 and 500μm. The in vitro dissolution behavior and bioactivity of the dual α/β-TCP were also probed in static and dynamic SBF for the first time. The results revealed that α/β-TCP scaffolds had good in vitro bioactivity, as the formation of bone-like apatite layers was induced on the scaffolds after mineralization in SBF. Moreover, dissolution rate of α/β-TCP bioceramics in dynamic environment was higher than that under static condition. Compared with monophasic TCP ceramics, these porous α/β-TCP bioceramics displayed a tailored dissolution rate proportionate to the TCP content (α and β) in the materials. Further, the degradation profile of porous α/β-TCP was well-described by Avrami equation. The porous dual α/β-TCP bioceramics with controllable degradation behavior hold great potential to be applied in bone tissue engineering as bone substitutes.
Sigma-Aldrich
Hydroxyapatite, powder, 5 μm
Sigma-Aldrich
Hydroxyapatite, powder, 10 μm
Sigma-Aldrich
Tricalcium phosphate, powder, 4 μm, ≥80 m2g
Sigma-Aldrich
Hydroxyapatite, aqueous paste, <50 nm, 30 wt. %
Sigma-Aldrich
Hydroxyapatite coating powder, ≥99% trace metals basis, ≤50 ppm(Pb)
Sigma-Aldrich
Hydroxyapatite, aqueous paste, <50 nm, 15 wt. %
Sigma-Aldrich
Hydroxyapatite, powder, 2.5 μm