Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, AnhydroBeads™, −10 mesh, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
7-Methylguanosine 5′-diphosphate sodium salt, ≥92.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, AnhydroBeads™, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-DCPS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Pricing and availability is not currently available.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Pricing and availability is not currently available.