- Magnetic nanoparticle-based theranostics.
ABSTRACT
This theme issue provides a timely collection of studies on magnetic nanoparticle-based imaging, bio-sensing, therapy and/or their combinations.
Sigma-Aldrich
Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), carboxylic acid functionalized, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL in H2O
Sigma-Aldrich
Iron oxide (II,III), nanoparticles, 5 nm avg. part. size (TEM), 5 mg/mL in chloroform
Sigma-Aldrich
Iron oxide (II,III), nanoparticles, 10 nm avg. part. size (TEM), 5 mg/mL in chloroform
Sigma-Aldrich
Iron oxide (II,III), nanoparticles, 10 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
Sigma-Aldrich
Iron oxide (II,III), nanoparticles, 30 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
Sigma-Aldrich
Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
Sigma-Aldrich
Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide (II,III), nanoparticles, 20 nm avg. part. size (TEM), 5 mg/mL in chloroform