Merck
Manganese-enhanced MRI of minimally gadolinium-enhancing breast tumors.
Journal of magnetic resonance imaging : JMRI (2014-03-05)
Tameshwar Ganesh, Reza Bayat Mokhtari, Mosa Alhamami, Herman Yeger, Hai-Ling Margaret Cheng
ABSTRACT
To investigate the potential of manganese (Mn)-enhanced MRI for sensitive detection and delineation of tumors that demonstrate little enhancement on Gd-DTPA. Eighteen nude rats bearing 1 to 2 cm in diameter orthotopic breast tumors (ZR75 and LM2) were imaged on a 3 Tesla (T) clinical scanner. Gd-DTPA was administered intravenously and MnCl2 subcutaneously, both at 0.05 mmol/kg. T1 -weighted imaging and T1 measurements were performed precontrast, 10 min post-Gd-DTPA, and 24 h post-MnCl2 . Tumors were excised and histologically assessed using H&E (composition and necrosis) and CD34 (vascularity). Most tumors (78%) demonstrated little enhancement (< 20% change in R1 ) on Gd-DTPA. MnCl2 administration achieved greater and more uniform enhancement throughout the tumor mass (i.e., not restricted to the tumor periphery), with R1 changing over 20% in 72% of tumors. MnCl2 -induced R1 changes compared with Gd-induced changes were significantly greater in both ZR75 (P < 0.01) and LM2 tumors (P < 0.05). Histology confirmed very low vascularity in both tumor models, and necrotic areas were well delineated only on Mn-enhanced MRI. Mn-enhanced MRI is a promising approach for detection of low-Gd-enhancing tumors.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ReagentPlus®, ≥99%