Merck
  • Analgesic topical capsaicinoid therapy increases somatostatin-like immunoreactivity in the human plasma.
Analgesic topical capsaicinoid therapy increases somatostatin-like immunoreactivity in the human plasma.
Neuropeptides (2014-12-03)
Katalin Horváth, Melinda Boros, Teréz Bagoly, Viktor Sándor, Ferenc Kilár, Agnes Kemény, Zsuzsanna Helyes, János Szolcsányi, Erika Pintér
ABSTRACT
The aim of the present study was to evaluate the therapeutic potential of local capsaicinoid (EMSPOMA(®) cream) treatment on chronic low back pain in patients with degenerative spine diseases and to investigate the possible mechanism of action of the therapy. The qualitative and quantitative analyses of capsaicinoids in EMSPOMA(®) cream were performed by liquid chromatography-tandem mass spectrometry (LC-MS/MS). In the clinical study 20 patients with degenerative spine diseases were involved in a self-controlled examination. During the 21 day therapy they received 30 min daily treatment with capsaicinoid (EMSPOMA(®)) cream to the lumbar region of the back. The pain (VASs, Oswestry Disability Index) and the mobility of the lumbar region of the spine (Schober's, Domján's L and R test) were detected at baseline and at the end of the 1st, 2nd and 3rd weeks. The plasma level of somatostatin-like immunoreactivity (SST-LI) was measured by radioimmunoassay (RIA) before and after the treatment on the first and the last day of the therapy. Nonivamide (0.01%) was identified as the only capsaicinoid molecule in the cream. In the clinical study the 21 day local nonivamide treatment reduced the pain sensation. Oswestry Disability Index decreased from 39 ± 3.9% to 32.5 ± 4.4%. VASs showed 37.29%-59.51% improvement. In the plasma level of SST-LI threefold elevation was observed after the first nonivamide treatment. We conclude that nonivamide treatment exerts analgesic action in chronic low back pain and causes the release of the antinociceptive and anti-inflammatory neuropeptide somatostatin which may play pivotal role in the pain-relieving effect.
Sigma-Aldrich
N-Vanillylnonanamide, ≥97%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Supelco
N-Vanillylnonanamide, analytical standard