Merck
  • Synthesis and pharmacological evaluation of polyfunctional benzimidazole-NSAID chimeric molecules combining anti-inflammatory, immunomodulatory and antioxidant activities.
Synthesis and pharmacological evaluation of polyfunctional benzimidazole-NSAID chimeric molecules combining anti-inflammatory, immunomodulatory and antioxidant activities.
Archives of pharmacal research (2013-11-06)
Yogita Bansal, Om Silakari
ABSTRACT
Polyfunctional compounds comprise a novel class of therapeutic agents for treatment of multifactorial diseases. The present study reports a series of benzimidazole-non-steroidal anti-inflammatory drugs (NSAIDs) conjugates (1-10) as novel polyfunctional compounds synthesized in the presence of orthophosphoric acid. The compounds were evaluated for anti-inflammatory (carageenan-induced paw edema model), immunomodulatory (direct haemagglutination test and carbon clearance index models), antioxidant (in vitro and in vivo) and for ulcerogenic effects. Each of the compound has retained the anti-inflammatory activity of the corresponding parent NSAID while exhibiting significantly reduced gastric ulcers. Additionally, the compounds are found to possess potent immunostimulatory and antioxidant activities. The compound 8 was maximally potent (antibody titre value 358.4 ± 140.21, carbon clearance index 0.053 ± 0.002 and antioxidant EC50 value 0.03 ± 0.006). These compounds, exhibiting such multiple pharmacological activities, can be taken as lead for the development of potent drugs for the treatment of chronic multifactorial diseases involving inflammation, immune system modulation and oxidative stress such as cancers. The Lipinski's parameters suggested the compounds to be bear drug like properties.
Sigma-Aldrich
4-Biphenylacetic acid, 98%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Mefenamic acid
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ibuprofen
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Mefenamic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Salicylic acid, ≥99%, FG
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Felbinac, European Pharmacopoeia (EP) Reference Standard
Mesalazine for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Mefenamic acid, analytical standard