Merck
  • Bcl-xL controls a switch between cell death modes during mitotic arrest.
Bcl-xL controls a switch between cell death modes during mitotic arrest.
Cell death & disease (2014-06-13)
N Bah, L Maillet, J Ryan, S Dubreil, F Gautier, A Letai, P Juin, S Barillé-Nion
ABSTRACT
Antimitotic agents such as microtubule inhibitors (paclitaxel) are widely used in cancer therapy while new agents blocking mitosis onset are currently in development. All these agents impose a prolonged mitotic arrest in cancer cells that relies on sustained activation of the spindle assembly checkpoint and may lead to subsequent cell death by incompletely understood molecular events. We have investigated the role played by anti-apoptotic Bcl-2 family members in the fate of mitotically arrested mammary tumor cells treated with paclitaxel, or depleted in Cdc20, the activator of the anaphase promoting complex. Under these conditions, a weak and delayed mitotic cell death occurs that is caspase- and Bax/Bak-independent. Moreover, BH3 profiling assays indicate that viable cells during mitotic arrest are primed to die by apoptosis and that Bcl-xL is required to maintain mitochondrial integrity. Consistently, Bcl-xL depletion, or treatment with its inhibitor ABT-737 (but not with the specific Bcl-2 inhibitor ABT-199), during mitotic arrest converts cell response to antimitotics to efficient caspase and Bax-dependent apoptosis. Apoptotic priming under conditions of mitotic arrest relies, at least in part, on the phosphorylation on serine 62 of Bcl-xL, which modulates its interaction with Bax and its sensitivity to ABT-737. The phospho-mimetic S62D-Bcl-xL mutant is indeed less efficient than the corresponding phospho-deficient S62A-Bcl-xL mutant in sequestrating Bax and in protecting cancer cells from mitotic cell death or yeast cells from Bax-induced growth inhibition. Our results provide a rationale for combining Bcl-xL targeting to antimitotic agents to improve clinical efficacy of antimitotic strategy in cancer therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Bcl2l1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nkx3-1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Bcl2
Sigma-Aldrich
MISSION® esiRNA, targeting human BCL2L1
Sigma-Aldrich
MISSION® esiRNA, targeting human BAK1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mcl1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cdc20
Sigma-Aldrich
MISSION® esiRNA, targeting human NKX3-1
Sigma-Aldrich
MISSION® esiRNA, targeting human BAX
Sigma-Aldrich
MISSION® esiRNA, targeting human MCL1
Sigma-Aldrich
MISSION® esiRNA, targeting human CDC20
Sigma-Aldrich
MISSION® esiRNA, targeting human BCL2
Sigma-Aldrich
Anti-BCL-XL (pS62) Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-phospho-Ser/Thr-Pro MPM-2 Antibody, FITC conjugate, Upstate®, from mouse