Merck
  • Basophil activation through ASGM1 stimulation triggers PAF release and anaphylaxis-like shock in mice.
Basophil activation through ASGM1 stimulation triggers PAF release and anaphylaxis-like shock in mice.
European journal of immunology (2014-04-30)
Yan Yang, Daling Li, Foad Katirai, Bin Zhang, Yong Xu, Ping Xiong, Feili Gong, Fang Zheng
ABSTRACT
Basophils have been shown to contribute to anaphylaxis through either an IgE-FcεRI-dependent pathway or an IgG-FcγR pathway. However, it remains largely unclear whether basophils can be activated to promote anaphylaxis via a non-FcR pathway as well. The glycolipid receptor ASGM1 (Asialoganglioside gangliotetraosylceramide), which has an exposed GalNAcβ1-4Gal moiety and serves as a receptor for pathogen associated molecular patterns such as flagellin, was recently found to be expressed on basophils. Here, we demonstrate that stimulation of basophils with anti-ASGM1 antibodies promotes platelet-activating factor (PAF) secretion in vitro and in vivo. Moreover, we found that ASGM1 stimulation triggers basophil- and PAF-dependent anaphylactic shock in pertussis toxin (PTX)-pretreated mice. Thus, ASGM1 has a crucial role in basophil activation and basophil-mediated anaphylaxis-like shock in mice, especially when the vascular permeability is increased by PTX treatment. Our findings describe a novel anaphylaxis-associated pathway that is antigen-, antibody-, and FcR-independent.
Sigma-Aldrich
Gadolinium(III) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Toluidine Blue, 8.74% (ZN (THEORY)), for microscopy (Hist., Vit.)
Supelco
Gadolinium(III) chloride solution, NMR reference standard, water 1 %, DSS 0.1 %, 0.1 mg/mL in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), Methanol-13C 0.1 % (99 atom % 13C), water 1 %, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), water 0.1 %, NMR tube size 6.5 mm × 8 in.
Supelco
Histamine, analytical standard
Sigma-Aldrich
Histamine, ≥97.0%