Merck
  • Quantitative analysis by GC-MS/MS of 18 aroma compounds related to oxidative off-flavor in wines.
Quantitative analysis by GC-MS/MS of 18 aroma compounds related to oxidative off-flavor in wines.
Journal of agricultural and food chemistry (2015-03-31)
Christine M Mayr, Dimitra L Capone, Kevin H Pardon, Cory A Black, Damian Pomeroy, I Leigh Francis
ABSTRACT
A quantitation method for 18 aroma compounds reported to contribute to "oxidative" flavor in wines was developed. The method allows quantitation of the (E)-2-alkenals ((E)-2-hexenal, (E)-2-heptenal, (E)-2-octenal, and (E)-2-nonenal), various Strecker aldehydes (methional, 2-phenylacetaldehyde, 3-methylbutanal, and 2-methylpropanal), aldehydes (furfural, 5-methylfurfural, hexanal, and benzaldehyde), furans (sotolon, furaneol, and homofuraneol), as well as alcohols (methionol, eugenol, and maltol) in the same analysis. The aldehydes were determined after derivatization directly in the wine with O-(2,3,4,5,6-pentafluorobenzyl)hydroxylamine hydrochloride; the formed oximes along with the underivatized aroma compounds were isolated by solid-phase extraction and analyzed by means of GC-MS/MS. The method was used to investigate the effect of different closures (synthetic closures, natural corks, and screw cap) on the formation of oxidation-related compounds in 14 year old white wine. Results showed a significant increase in the concentration of some of the monitored compounds in the wine, particularly methional, 2-phenylacetaldehyde, and 3-methylbutanal.
Sigma-Aldrich
Maltol, ≥99.0%, FCC, FG
Sigma-Aldrich
Maltol, natural, FG
Supelco
Maltol, analytical standard
Sigma-Aldrich
3-Hydroxy-2-methyl-4-pyrone, 99%