Merck
  • Evidence for a hexaheteromeric methylenetetrahydrofolate reductase in Moorella thermoacetica.
Evidence for a hexaheteromeric methylenetetrahydrofolate reductase in Moorella thermoacetica.
Journal of bacteriology (2014-07-09)
Johanna Mock, Shuning Wang, Haiyan Huang, Jörg Kahnt, Rudolf K Thauer
ABSTRACT
Moorella thermoacetica can grow with H₂ and CO₂, forming acetic acid from 2 CO₂ via the Wood-Ljungdahl pathway. All enzymes involved in this pathway have been characterized to date, except for methylenetetrahydrofolate reductase (MetF). We report here that the M. thermoacetica gene that putatively encodes this enzyme, metF, is part of a transcription unit also containing the genes hdrCBA, mvhD, and metV. MetF copurified with the other five proteins encoded in the unit in a hexaheteromeric complex with an apparent molecular mass in the 320-kDa range. The 40-fold-enriched preparation contained per mg protein 3.1 nmol flavin adenine dinucleotide (FAD), 3.4 nmol flavin mononucleotide (FMN), and 110 nmol iron, almost as predicted from the primary structure of the six subunits. It catalyzed the reduction of methylenetetrahydrofolate with reduced benzyl viologen but not with NAD(P)H in either the absence or presence of oxidized ferredoxin. It also catalyzed the reversible reduction of benzyl viologen with NADH (diaphorase activity). Heterologous expression of the metF gene in Escherichia coli revealed that the subunit MetF contains one FMN rather than FAD. MetF exhibited 70-fold-higher methylenetetrahydrofolate reductase activity with benzyl viologen when produced together with MetV, which in part shows sequence similarity to MetF. Heterologously produced HdrA contained 2 FADs and had NAD-specific diaphorase activity. Our results suggested that the physiological electron donor for methylenetetrahydrofolate reduction in M. thermoacetica is NADH and that the exergonic reduction of methylenetetrahydrofolate with NADH is coupled via flavin-based electron bifurcation with the endergonic reduction of an electron acceptor, whose identity remains unknown.
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, suitable for electrophoresis, suitable for acrylamide photopolymerization, ≥70%, powder
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, 73-79% (fluorimetric)
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, synthetic, ≥70% (HPLC)
Adenine, European Pharmacopoeia (EP) Reference Standard
Supelco
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
Riboflavin 5′-phosphate sodium salt hydrate, meets USP testing specifications