Merck
  • Tumour-suppressive microRNA-24-1 inhibits cancer cell proliferation through targeting FOXM1 in bladder cancer.
Tumour-suppressive microRNA-24-1 inhibits cancer cell proliferation through targeting FOXM1 in bladder cancer.
FEBS letters (2014-07-08)
Satoru Inoguchi, Naohiko Seki, Takeshi Chiyomaru, Tomoaki Ishihara, Ryosuke Matsushita, Hiroko Mataki, Toshihiko Itesako, Shuichi Tatarano, Hirofumi Yoshino, Yusuke Goto, Rika Nishikawa, Masayuki Nakagawa, Hideki Enokida
ABSTRACT
Here, we found that microRNA-24-1 (miR-24-1) is significantly reduced in bladder cancer (BC) tissues, suggesting that it functions as a tumour suppressor. Restoration of mature miR-24-1 inhibits cancer cell proliferation and induces apoptosis. Forkhead box protein M1 (FOXM1) is a direct target gene of miR-24-1, as shown by genome-wide gene expression analysis and luciferase reporter assay. Overexpressed FOXM1 is confirmed in BC clinical specimens, and silencing of FOXM1 induces apoptosis in cancer cell lines. Our data demonstrate that the miR-24-1-FOXM1 axis contributes to cancer cell proliferation in BC, and elucidation of downstream signalling will provide new insights into the molecular mechanisms of BC oncogenesis.
Sigma-Aldrich
MISSION® esiRNA, targeting human FOXM1
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
SAFC
BIS-TRIS
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
SAFC
BIS-TRIS
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Foxm1
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse